siRNA Details
| Gene Symbol: 4921509J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014853.1 | - | 1 | 423 | ||
| AK054449.1 | - | 1 | 1606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921509J17Rik| Gene Symbol: 4921509J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014853.1 | - | 1 | 423 | ||
| AK054449.1 | - | 1 | 1606 | |||