siRNA Details
| Gene Symbol: 2900002H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019300.1 | - | 1 | 177 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712438.1 | - | 1 | 175 | Chr. 1 - 38322228 - 38322499 [-] on Build GRCm38 | |

2900002H24Rik
Genomic Map