siRNA Details
Gene Symbol: Gm36426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033777.1 | - | 2, 3 | 1415 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV380962.2 | - | 2, 3 | 291 | Chr. 13 - 70676720 - 70680811 [-] on Build GRCm38 |
