siRNA Details
| Gene Symbol: Gm36426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033777.1 | - | 2 | 1347 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV380962.2 | - | 2 | 223 | Chr. 13 - 70676720 - 70680811 [-] on Build GRCm38 | |

Gm36426
Genomic Map