siRNA Details
| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082774.1 | - | 1 | 820 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB399121.2 | - | 1 | 238 | Chr. 11 - 57726254 - 57726911 [+] on Build GRCm38 | |

Galnt10
Genomic Map