siRNA Details
| Gene Symbol: 9030625G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110446.1 | 5 | 580 | |||
| GenBank mRNA | AK018576.1 | - | 5 | 581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030625G05Rik| Gene Symbol: 9030625G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110446.1 | 5 | 580 | |||
| GenBank mRNA | AK018576.1 | - | 5 | 581 | ||