siRNA Details
| Gene Symbol: Gm12596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105031.1 | 3 | 357 | |||
| GenBank mRNA | AK143878.1 | - | 3 | 358 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY732728.1 | - | 3 | 358 | Chr. 4 - 59260051 - 59263244 [+] on Build GRCm38 | |

Gm12596
Genomic Map