siRNA Details
| Gene Symbol: 1700063D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040392.1 | 1 | 84 | |||
| GenBank mRNA | AK018871.1 | - | 1 | 81 | ||
| BC048584.1 | - | 1 | 51 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700063D05Rik| Gene Symbol: 1700063D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040392.1 | 1 | 84 | |||
| GenBank mRNA | AK018871.1 | - | 1 | 81 | ||
| BC048584.1 | - | 1 | 51 | |||