siRNA Details
| Gene Symbol: 1700065F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018878.1 | - | 1 | 45 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706880.1 | - | 1 | 44 | Chr. 2 - 85086620 - 85086981 [+] on Build GRCm38 | |

1700065F09Rik
Genomic Map