siRNA Details
Gene Symbol: 4930517O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015818.1 | - | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715362.1 | - | 1 | 233 | Chr. 14 - 100213189 - 100246223 [+] on Build GRCm38 |
