siRNA Details
| Gene Symbol: 4930517O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015818.1 | - | 962 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930517O19Rik| Gene Symbol: 4930517O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015818.1 | - | 962 | |||