siRNA Details
Gene Symbol: 4930517O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015818.1 | - | 729 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615807.1 | - | 5 | 533 | Chr. 14 - 100213142 - 100245339 [+] on Build GRCm38 |
