siRNA Details
| Gene Symbol: 4930517J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015815.1 | - | 1 | 28 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715357.1 | - | 1 | 25 | Chr. 4 - 44669333 - 44670050 [+] on Build GRCm38 | |

4930517J16Rik
Genomic Map