siRNA Details
| Gene Symbol: 2210416O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045499.1 | 1 | 47 | |||
| GenBank mRNA | AK008958.1 | - | 1 | 48 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708845.1 | - | 1 | 48 | Chr. 11 - 88098058 - 88099419 [+] on Build GRCm38 | |

2210416O15Rik
Genomic Map