siRNA Details
| Gene Symbol: 2210416O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045499.1 | 4 | 1403 | |||
| GenBank mRNA | AK008958.1 | - | 4 | 1400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210416O15Rik| Gene Symbol: 2210416O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045499.1 | 4 | 1403 | |||
| GenBank mRNA | AK008958.1 | - | 4 | 1400 | ||