siRNA Details
| Gene Symbol: 1010001N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105022.1 | 4 | 492 | |||
| GenBank mRNA | AK003136.1 | - | 4 | 493 | ||
| AK033147.1 | - | 2 | 887 | |||
| CF382894.1 | - | 1 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1010001N08Rik| Gene Symbol: 1010001N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105022.1 | 4 | 492 | |||
| GenBank mRNA | AK003136.1 | - | 4 | 493 | ||
| AK033147.1 | - | 2 | 887 | |||
| CF382894.1 | - | 1 | 203 | |||