siRNA Details
| Gene Symbol: 1010001N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105022.1 | 2 | 185 | |||
| NR_105023.1 | 2 | 185 | ||||
| GenBank mRNA | AK003136.1 | - | 2 | 186 | ||
| AK033147.1 | - | 1 | 213 | |||
| CK332479.1 | - | 148 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1010001N08Rik| Gene Symbol: 1010001N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105022.1 | 2 | 185 | |||
| NR_105023.1 | 2 | 185 | ||||
| GenBank mRNA | AK003136.1 | - | 2 | 186 | ||
| AK033147.1 | - | 1 | 213 | |||
| CK332479.1 | - | 148 | ||||