siRNA Details
| Gene Symbol: Gm28515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136272.1 | - | 2 | 2466 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB064740.2 | - | 2 | 632 | Chr. 8 - 91907565 - 91911235 [-] on Build GRCm38 | |

Gm28515
Genomic Map