siRNA Details
| Gene Symbol: Slc25a51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020275.1 | - | 2 | 638 | ||
| AK172225.1 | - | 1 | 1199 | |||
| BC026435.1 | - | 1 | 695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a51| Gene Symbol: Slc25a51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020275.1 | - | 2 | 638 | ||
| AK172225.1 | - | 1 | 1199 | |||
| BC026435.1 | - | 1 | 695 | |||