siRNA Details
| Gene Symbol: Gm33466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139694.1 | - | 3 | 1601 | ||
| Gene Symbol: Gm11783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI838599.1 | - | 1 | 271 | ||

Gm33466
Genomic Map