siRNA Details
| Gene Symbol: Gm15473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144868.1 | - | 2 | 448 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY734531.1 | - | 2 | 448 | Chr. 6 - 17059285 - 17064996 [-] on Build GRCm38 | |

Gm15473
Genomic Map