siRNA Details
| Gene Symbol: Gm15496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135877.1 | - | 4 | 1900 | ||
| AK162138.1 | - | 4 | 1900 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15496| Gene Symbol: Gm15496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135877.1 | - | 4 | 1900 | ||
| AK162138.1 | - | 4 | 1900 | |||