siRNA Details
| Gene Symbol: Gm15496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135877.1 | - | 4 | 681 | ||
| AK162138.1 | - | 4 | 681 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV658648.1 | - | 3 | 631 | Chr. 3 - 31311019 - 31315297 [+] on Build GRCm38 | |

Gm15496
Genomic Map