siRNA Details
| Gene Symbol: Gm10634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077175.1 | - | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10634| Gene Symbol: Gm10634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077175.1 | - | 614 | |||