siRNA Details
| Gene Symbol: Gm10634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077175.1 | - | 1645 | |||
| Gene Symbol: 4930579C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029815.1 | - | 13 | 2733 | ||

Gm10634
Genomic Map