siRNA Details
| Gene Symbol: Gm29927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039399.1 | - | 2 | 343 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB633328.1 | - | 2 | 344 | Chr. 13 - 102566149 - 102590285 [-] on Build GRCm38 | |

Gm29927
Genomic Map