siRNA Details
| Gene Symbol: 9030408N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018491.1 | - | 1 | 568 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720664.1 | - | 1 | 568 | Chr. 15 - 34665287 - 34666052 [-] on Build GRCm38 | |

9030408N04Rik
Genomic Map