siRNA Details
| Gene Symbol: 5830426K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017949.1 | - | 1 | 878 | ||
| AK041807.1 | - | 1 | 1922 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830426K05Rik| Gene Symbol: 5830426K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017949.1 | - | 1 | 878 | ||
| AK041807.1 | - | 1 | 1922 | |||