siRNA Details
| Gene Symbol: 5830426K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017949.1 | - | 1 | 234 | ||
| AK041807.1 | - | 1 | 1276 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717837.1 | - | 1 | 234 | Chr. 11 - 87341989 - 87342809 [-] on Build GRCm38 | |

5830426K05Rik
Genomic Map