siRNA Details
| Gene Symbol: 1810059H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007907.1 | - | 2 | 640 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB805886.1 | - | 1 | 334 | Chr. 5 - 148958598 - 148958981 [-] on Build GRCm38 | |

1810059H22Rik
Genomic Map