siRNA Details
| Gene Symbol: 1810058N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007900.1 | - | 1 | 130 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707903.1 | - | 1 | 126 | Chr. 19 - 5561056 - 5561676 [+] on Build GRCm38 | |

1810058N15Rik
Genomic Map