siRNA Details
| Gene Symbol: 1810059C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007903.1 | - | 3 | 313 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707906.1 | - | 3 | 313 | Chr. 2 - 18054193 - 18056613 [-] on Build GRCm38 | |

1810059C17Rik
Genomic Map