siRNA Details
Gene Symbol: 1810059C17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007903.1 | - | 3 | 251 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707906.1 | - | 3 | 251 | Chr. 2 - 18054193 - 18056613 [-] on Build GRCm38 |
