siRNA Details
| Gene Symbol: Tmem182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010233.1 | - | 1 | 1192 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA792460.1 | - | 1 | 46 | Chr. 1 - 40856352 - 40856828 [+] on Build GRCm38 | |

Tmem182
Genomic Map