siRNA Details
| Gene Symbol: C230021G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045502.1 | - | 2 | 2138 | ||
| AK082191.1 | - | 2 | 1937 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C230021G24Rik| Gene Symbol: C230021G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045502.1 | - | 2 | 2138 | ||
| AK082191.1 | - | 2 | 1937 | |||