siRNA Details
| Gene Symbol: A930011O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040709.1 | 4 | 3538 | |||
| GenBank mRNA | AK032410.1 | - | 1 | 1175 | ||
| BC030402.1 | - | 2 | 3323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930011O12Rik| Gene Symbol: A930011O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040709.1 | 4 | 3538 | |||
| GenBank mRNA | AK032410.1 | - | 1 | 1175 | ||
| BC030402.1 | - | 2 | 3323 | |||