siRNA Details
| Gene Symbol: A930011O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040709.1 | 4 | 3246 | |||
| GenBank mRNA | AK032410.1 | - | 1 | 883 | ||
| BC030402.1 | - | 2 | 3031 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU505303.1 | - | 1 | 63 | Chr. 14 - 64593106 - 64593958 [+] on Build GRCm38 | |

A930011O12Rik
Genomic Map