siRNA Details
| Gene Symbol: 1810028F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007631.1 | - | 1 | 935 | ||
| AK007949.1 | - | 1 | 606 | |||
| AV053782.1 | - | 1 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810028F09Rik| Gene Symbol: 1810028F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007631.1 | - | 1 | 935 | ||
| AK007949.1 | - | 1 | 606 | |||
| AV053782.1 | - | 1 | 181 | |||