siRNA Details
| Gene Symbol: 5830428M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038060.1 | 5 | 952 | |||
| NR_038061.1 | 5 | 946 | ||||
| GenBank mRNA | AK039829.1 | - | 5 | 949 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830428M24Rik| Gene Symbol: 5830428M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038060.1 | 5 | 952 | |||
| NR_038061.1 | 5 | 946 | ||||
| GenBank mRNA | AK039829.1 | - | 5 | 949 | ||