siRNA Details
| Gene Symbol: A430010J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039826.1 | - | 354 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB634278.1 | - | 3, 4 | 354 | Chr. 6 - 67115673 - 67167020 [+] on Build GRCm38 | |

A430010J10Rik
Genomic Map