siRNA Details
| Gene Symbol: Gm3896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072643.1 | - | 2 | 2660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3896| Gene Symbol: Gm3896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072643.1 | - | 2 | 2660 | ||