siRNA Details
| Gene Symbol: 3100002H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013917.1 | - | 1 | 735 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3100002H20Rik| Gene Symbol: 3100002H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013917.1 | - | 1 | 735 | ||