siRNA Details
| Gene Symbol: 3100002H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013917.1 | - | 1 | 422 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712955.1 | - | 1 | 419 | Chr. 15 - 72786179 - 72787083 [-] on Build GRCm38 | |

3100002H20Rik
Genomic Map