siRNA Details
| Gene Symbol: A230045G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138253.1 | - | 5 | 932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230045G11Rik| Gene Symbol: A230045G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138253.1 | - | 5 | 932 | ||