siRNA Details
| Gene Symbol: A230045G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138253.1 | - | 5 | 1440 | ||
| Gene Symbol: Gpr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ175263.1 | - | 1 | 361 | ||

A230045G11Rik
Genomic Map