siRNA Details
| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047922.1 | - | 1 | 626 | ||
| AK079567.1 | - | 29 | ||||
| AK159465.1 | - | 1 | 3081 | |||
| BC100372.1 | - | 1 | 1362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxo3| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047922.1 | - | 1 | 626 | ||
| AK079567.1 | - | 29 | ||||
| AK159465.1 | - | 1 | 3081 | |||
| BC100372.1 | - | 1 | 1362 | |||