siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135177.1 | - | 1 | 533 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ050984.1 | - | 1 | 533 | Chr. 11 - 115926962 - 115927605 [-] on Build GRCm38 | |

Recql5
Genomic Map