siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135177.1 | - | 1 | 1346 | ||
| Gene Symbol: Gm9978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081416.1 | - | 1 | 2623 | ||

Recql5
Genomic Map