siRNA Details
| Gene Symbol: Gm13480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044108.1 | - | 3 | 582 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB642037.1 | - | 3 | 581 | Chr. 2 - 49806363 - 49812948 [-] on Build GRCm38 | |

Gm13480
Genomic Map