siRNA Details
| Gene Symbol: 9530049O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020601.1 | - | 1 | 476 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722137.1 | - | 1 | 476 | Chr. 5 - 31957067 - 31957769 [+] on Build GRCm38 | |

9530049O05Rik
Genomic Map